SureTight™ Sterile Sample Cup with Polyethylene Screw Cap
Supplier: Hardy Diagnostics
243810
77497-250PK 497.11 USD
77497-250
SureTight™ Sterile Sample Cup with Polyethylene Screw Cap
Sample Containers Sample Cups for Analyzers
The lid is designed to securely lock in the sample. When you use the SureTight™, you can be confident that your sample is safely contained in the cup and will remain that way throughout transport to the lab.
  • Tamper-proof sterility tab ensures sample integrity
  • The cups feature a wide 53 mm opening for larger samples
  • Ergonomic design features lids with ridges that allow for easy opening
  • Stackable and interlocking for efficient use of space
  • Economical-by far, SureTight™ represents the best value
  • The SureTight™ sampling cups are tough, leakproof, sterile