2,2,2-trifluoro-1-(5-methylbenzo[b]thiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306073-1G 306073-500MG 306073-5G
77468-676EA 415.09 USD
77468-676 77481-360 77491-742
2,2,2-trifluoro-1-(5-methylbenzo[b]thiophen-2-yl)ethanone ≥95%
2,2,2-trifluoro-1-(5-methylbenzo[b]thiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₁₁H₇F₃OS
MW: 244.2328 g/mol
Storage Temperature: Ambient
CAS Number: 3002426-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR