1-(3-Bromophenyl)octan-1-ol ≥97%

Supplier: AOBChem USA

Warning

154620-1G 154620-500MG 154620-5G 154620-25G
77465-108EA 415.09 USD
77465-108 77477-876 77488-014 77498-704
1-(3-Bromophenyl)octan-1-ol ≥97%
1-(3-Bromophenyl)octan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₁BrO
MW: 285.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18352306
CAS Number: 162359-13-9
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR