(2-Bromo-6-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

143592-1G 143592-500MG 143592-5G 143592-25G
77464-898EA 415.09 USD
77464-898 77477-674 77487-794 77498-510
(2-Bromo-6-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
(2-Bromo-6-(trifluoromethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₆BrF₃OS
MW: 287.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34836708
CAS Number: 2167829-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR