1-(3-Chlorophenyl)cyclooctanol ≥95%

Supplier: AOBChem USA

Warning

129546-1G 129546-500MG 129546-5G 129546-25G
77464-536EA 415.09 USD
77464-536 77477-322 77487-424 77498-218
1-(3-Chlorophenyl)cyclooctanol ≥95%
1-(3-Chlorophenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₄H₁₉ClO
MW: 238.753 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828150
CAS Number: 2921849-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR