Bravo Disposable Pipette Tips
Supplier: Agilent Technologies
10734-202 10734-212 10734-302 10734-312 11484-202 11484-212 11484-302 11484-312 11484-322 19477-002 19477-012 19477-022 19477-032 19477-072 19477-082 21740-202 19133-102 19133-112 19133-142 19133-202 19133-212 19134-002 19134-012 19134-022 21740-212 21750-202 21750-212
77463-608PK 4037.12 USD
77463-608 77463-610 77463-612 77463-614 77463-616 77463-618 77463-620 77463-622 77463-624 77463-646 77463-648 77463-650 77463-652 77463-654 77463-656 77463-704 77463-630 77463-632 77463-634 77463-636 77463-638 77463-640 77463-642 77463-644 77463-706 77463-708 77463-710
Bravo Disposable Pipette Tips
Pipette Tips
Certified on Bravo and AssayMAP for accuracy and precision.
  • Individual lot certified to meet thorough QC standards
  • Certified free of detectable DNase and RNase activity
  • Certified free of endotoxin (pyrogen) contamination
  • Certified free of adenosine triphosphate (ATP)
  • Certified free of human DNA;Certified free of protease activity
  • Certified free of heavy metals
  • Certified as sterile with a sterility assurance level (SAL) of 10-6
  • Innovative processing and packaging reduce static buildup and improve deck space utilization
Agilent offers disposable pipetteMore Product Information