NIGHTSEA® Stereo Microscope Fluorescence Adapters (SFA), Full System
Supplier: Electron Microscopy Sciences
SFA-RB SFA-CY SFA-GR SFA-LFS-RB SFA-LFS-CY SFA-LFS-GR SFA-RB-GO-DIM SFA-CY-DIM SFA-CY-PULSE SFA-GR-DIM SFA-GR-PULSE SFA-LFS-RB-GO SFA-LFS-UV SFA-LFS-VI SFA-RB-DIM SFA-RB-GO-PULSE SFA-RB-PULSE SFA-UV SFA-UV-DIM SFA-UV-PULSE SFA-VI SFA-VI-DIM SFA-VI-PULSE
102773-514EA 2331.28 USD
102773-514 102773-516 102773-518 102773-520 102773-522 102773-524 MSPP-SFARBGODI 77463-306 77463-308 77463-314 77463-316 77463-318 77463-320 77463-322 77463-328 77463-330 77463-332 77463-338 77463-340 77463-342 77463-344 77463-346 77463-348
NIGHTSEA® Stereo Microscope Fluorescence Adapters (SFA), Full System
Lights Fluorescent Lights, Lamps and Bulbs
The NIGHTSEA® stereo microscope fluorescence adapter system adapts almost all stereo microscopes for fluorescence viewing without needing modifications.
  • Lamp base with power supply and international plug set
  • Light head: ultraviolet, violet, royal blue, cyan, or green
  • Barrier filter
  • Viewing shield
  • Padded travel case
The modular design easily switches between different excitation/emission combinations to work with a variety of fluorescent proteins and other fluorophores.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable