NEXTFLEX™ Unique Dual Index Barcodes (Set of 1536)
Supplier: REVVITY HEALTH SCIENCES, INC.
NOVA-534100 NOVA-634108 NOVA-634109 NOVA-634110 NOVA-634111 NOVA-634112 NOVA-634113 NOVA-634114 NOVA-634115 NOVA-634116 NOVA-634100 NOVA-634100-EVAL16 NOVA-634100-EVAL48 NOVA-634101 NOVA-634102 NOVA-634103 NOVA-534101 NOVA-534102 NOVA-534103 NOVA-534104 NOVA-634104 NOVA-634105 NOVA-634106 NOVA-634107
77448-206EA 20857.5 USD
77448-206 77448-248 77448-250 77448-252 77448-254 77448-256 77448-258 77448-260 77448-262 77448-264 77448-228 77448-230 77448-232 77448-234 77448-236 77448-238 77448-208 77448-210 77448-212 77448-214 77448-240 77448-242 77448-244 77448-246
NEXTFLEX™ Unique Dual Index Barcodes (Set of 1536)
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX® UDI Barcodes (1 to 1536) are 1536 unique dual index adapters with 10 nt indexes for multiplexing of NGS libraries compatible with Illumina® sequencers.
  • 384 UDI adapters for mitigating sample misassignment due to index hopping
  • Unique dual index adapters for multiplexing up to 384 libraries for Illumina® sequencing platforms
  • Mitigate index hopping or spread of signal that can occur during sequencing
  • Each lot of the NEXTFLEX® unique dual index barcodes is tested for index purity by sequencing
  • Compatible with both paired-end and single-read sequencing
  • Compatible with the NEXTFLEX® universal blockers
The NEXTFLEX unique dual index barcodes utilize two unique indexes on each UDI adapter, with a hamming distance of at least three throughout the entire series to permit error correction.
Each NEXTFLEX unique dual index barcode comprises the entire flow cell binding site, sequencing primer binding site and index sequences, thereby making PCR enrichment optional. Patterned flow cells, such as those found on the Illumina NovaSeq®, HiSeq® 3000/4000, and HiSeq X platforms, have been known to suffer from increased rates of sample mis-assignment during sequencing.
The use of adapters with unique dual indices improves data integrity by correctly removing mis-assigned reads. Every lot is tested for index purity and functionality by sequencing. The design of the NEXTFLEX unique dual index barcodes allow a broad range of multiplexing - users can multiplex anywhere between two samples and 1536 samples in a single sequencing run.
Certificaciones: Manufactured in ISO 9001 Facility.
Precaución: For research use only. Not for use in diagnostic procedures.