Anti-SLC23A3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-20881R
77439-330EA 380.63 USD
77439-330
Anti-SLC23A3 Rabbit Polyclonal Antibody
Antibodies
SLC23A3 (Solute Carrier Family 23, Member 3) is a Protein Coding gene. Diseases associated with SLC23A3 include epilepsy, generalized, with febrile seizures plus, type 5. GO annotations related to this gene include transporter activity. An important paralog of this gene is SLC23A1.

Type: Primary
Antigen: SLC23A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR