Advanced Indicator for Industial Applications
Supplier: Ohaus
30126959
77434-816EA 1262.31 USD
77434-816
Advanced Indicator for Industial Applications
Balances and Scales
Advanced performance stainless steel indicator with external device control capabilities.
With a 50000 displayed resolution and the ability to support up to ten load cells, T72XW was designed for heavy-duty industrial applications such as vehicle weighing. Two option slots are available to enable a wide range of connectivity interfaces. Control external devices using the discrete I/O interface and transfer data through the serial, USB or Ethernet ports. T72XW is very easy to set up and navigate - featuring a vivid graphic display with on-screen prompts and an alphanumeric keypad withMore Product Information