5-Fluoro-o-toluidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline

PC3822-100G PC3822-25G PC3822-500G
77414-928EA 27.5 USD
77414-928 77414-930 77414-932
5-Fluoro-o-toluidine 98%
5-Fluoro-o-toluidine
Formula: C₇H₈FN
MW: 125.15
Boiling Pt: 98…100 °C/15 mmHg
Melting Pt: 37…40 °C
Density: 1.13
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007764
CAS Number: 367-29-3
EINECS: 206-689-6
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR