6-Fluoro-o-toluidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Fluoro-6-methylaniline

PC1391-100G PC1391-25G PC1391-5G
77413-140EA 154.86 USD
77413-140 77413-142 77413-144
6-Fluoro-o-toluidine 97%
6-Fluoro-o-toluidine
Formula: C₇H₈FN
MW: 125.1435 g/mol
Boiling Pt: 88…89 °C/13 mmHg
Density: 1.082 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658252
CAS Number: 443-89-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR