4,4,4-Trifluorobutane-1-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC01483-250MG
77412-412EA 137.54 USD
77412-412
4,4,4-Trifluorobutane-1-sulfonyl chloride
4,4,4-Trifluorobutane-1-sulfonyl chloride
Formula: C₄H₆ClF₃O₂S
MW: 210.602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657959
CAS Number: 212190-25-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR