Benzophenone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

OR8243-1KG OR8243-500G
77411-380EA 103.92 USD
77411-380 77411-382
Benzophenone
Benzophenone
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305 °C
Melting Pt: 47…51 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9+EHS III
Merck Index: 13,01099

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR