2-Bromo-4-chloro-5-iodoanisole

Supplier: APOLLO SCIENTIFIC
Warning

OR400853-1G OR400853-5G
77409-490EA 274.06 USD
77409-490 77409-492
2-Bromo-4-chloro-5-iodoanisole
2-Bromo-4-chloro-5-iodoanisole
Formula: C₇H₅BrClIO
Melting Pt: 80…83 °C
MDL Number: MFCD11845921
CAS Number: 1160574-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR