4-(2-Hydroxy-3-morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(Morpholin-1-yl)-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propan-2-ol

OR3510-100MG
77409-304EA 83.54 USD
77409-304
4-(2-Hydroxy-3-morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester 97%
4-(2-Hydroxy-3-morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester
Formula: C₁₉H₃₀BNO₅
MW: 363.256 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258653
CAS Number: 756520-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR