N'-Hydroxy-2-pyrazinecarboximidamide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N'-Hydroxypyrazine-2-carboximidamide, Pyrazine-2-amidoxime

OR3401-250MG
77409-246EA 20.26 USD
77409-246
N'-Hydroxy-2-pyrazinecarboximidamide 97%
N'-Hydroxy-2-pyrazinecarboximidamide
Formula: C₅H₆N₄O
MW: 138.13 g/mol
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD02258397
CAS Number: 51285-05-3
UN: 2811
ADR: 6.1,III
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR