6-Bromobenzo[c]isothiazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Bromo-2,1-benzothiazole

OR16643-250MG
77405-814EA 571.56 USD
77405-814
6-Bromobenzo[c]isothiazole 95%
6-Bromobenzo[c]isothiazole
Formula: C₇H₄BrNS
MW: 214.082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176692
CAS Number: 20712-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR