2,6-Diaminopyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR13639-100G OR13639-500G
77404-694EA 96.78 USD
77404-694 77404-696
2,6-Diaminopyridine
2,6-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 285
Melting Pt: 117…122 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006329
CAS Number: 141-86-6
EINECS: 205-507-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR