Boc-3-amino-3-(3-bromophenyl)propionic acid

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-(3-Bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoic acid, 3-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

OR110967-1G
77403-600EA 93.74 USD
77403-600
Boc-3-amino-3-(3-bromophenyl)propionic acid
Boc-3-amino-3-(3-bromophenyl)propionic acid
Formula: C₁₄H₁₈BrNO₄
MW: 344.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02090700
CAS Number: 284493-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR