Ethoxydiphenylphosphine 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Diphenylphosphinous Acid Ethyl Ester

OR10812-100G
77402-934EA 103.92 USD
77402-934
Ethoxydiphenylphosphine 98%
Ethoxydiphenylphosphine
Formula: C₁₄H₁₅OP
MW: 230.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009087
CAS Number: 719-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR