Fold and Snap Tube Racks, 21 mm, 40-Place
Supplier: VWR International
HS243071BV HS243071RV HS243071WV HS243071YV
77395-400CS 577.09 USD
77395-400 77395-402 77395-404 77395-406
Fold and Snap Tube Racks, 21 mm, 40-Place
Racks Tube Racks
Economical durable rack.
  • Polypropylene racks ship flat in one piece
  • Easy-to-assemble, just fold and snap together
  • Stable design can be used in water baths
  • Open design allows samples to be incubated at the same temperature
  • Color code for easy identification
  • Easy to assemble, simply fold and snap together
The Fold and Snap Test tube rack is economical and durable.
Order Now
SPECIFICATIONS
Environmentally Preferable