Double Mega Rack for 10 to 13 mm Tubes
Supplier: VWR International
120112V 120113V
77395-306CS 241.25 USD
77395-306 77395-308
Double Mega Rack for 10 to 13 mm Tubes
Racks Tube Racks
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage.
  • Sturdy, material for easy and secure transport of blood collection and other tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack eliminating the need for multiple racks
The Mega Rack® high-capacity storage rack is great for high throughput labs requiring large sample storage.
Order Now
SPECIFICATIONS
Environmentally Preferable