Stilla® naica® IQ/OQ Kit
Supplier: STILLA TECHNOLOGIES, INC.
R30001
77392-294EA 489.6 USD
77392-294
Stilla® naica® IQ/OQ Kit
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Routine Operational Qualification (OQ) procedures of the naica® system and Nio™+ can be performed using the naica® IQ/OQ Kit.
  • Ready-to-use OQ solution
  • 24 reactions on the sapphire chip (Two full OQ of the naica® system)
  • 96 ruby chip reactions (1 full OQ of the Nio™+)
The naica® IQ/OQ Kit is a ready-to-use OQ solution to closely monitor equipment compliance to operating specifications. Use the naica® IQ/OQ Kit to qualify the naica® system and Nio™+ on a periodic basis in accordance with established operating procedures (SOPs). Experience the streamlined routine OQ solution with the naica® IQ/OQ Kit for a wide range of key nucleic detection and quantification applications, validated for use on all Stilla Technologies dPCR systems.
Información de suministro: Dry-ice shipment.
Precaución: Store at −20 ±5 °C. For research use only. Not for use in diagnostic procedures.