Stilla® Ruby Chip
Supplier: STILLA TECHNOLOGIES, INC.
C16011
77392-266PK 491.88 USD
77392-266
Stilla® Ruby Chip
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The Ruby Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single fully automation-compatible consumable.
  • Capacity: 16 samples/chip
  • Input volume: 5 µl/chamber
  • Droplets per sample: Up to 17000
  • Dynamic range: ∼4.7 log
  • Fully compatible with integration to automated liquid handling equipment
For higher throughput applications, the Ruby Chip offers the analysis of up to 48 samples per run for your digital PCR workflows. After sample preparation, just a single pipetting step is required before the chip is loaded onto either the naica® system or the Nio™+ for partitioning, PCR amplification, and sample read-out. The Ruby Chip is also fully compatible with integration to automated liquid handling equipment.