4-((cyclopropylmethyl)thio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

89970-500MG 89970-1G 89970-5G
77335-090EA 321.36 USD
77335-090 77359-176 77383-132
4-((cyclopropylmethyl)thio)benzaldehyde ≥97%
4-((cyclopropylmethyl)thio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂OS
MW: 192.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24686210
CAS Number: 917382-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR