3-Fluoro-4-isopropoxy-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

49652-500MG 49652-1G 49652-5G
77336-392EA 387.28 USD
77336-392 77351-540 77375-816
3-Fluoro-4-isopropoxy-2-(methylthio)benzoic acid ≥97%
3-Fluoro-4-isopropoxy-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃S
MW: 244.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564140
CAS Number: 2734775-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR