4-Fluoro-3-isopropoxy-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51764-500MG 51764-1G 51764-5G
77328-538EA 347.11 USD
77328-538 77352-130 77376-412
4-Fluoro-3-isopropoxy-5-(methylthio)benzoic acid ≥97%
4-Fluoro-3-isopropoxy-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃S
MW: 244.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902972
CAS Number: 2586127-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR