1-(4-Chlorophenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

64346-500MG 64346-1G 64346-5G 64346-25G
77332-158EA 169.95 USD
77332-158 77346-358 77368-222 77389-546
1-(4-Chlorophenyl)cyclohexanol ≥97%
1-(4-Chlorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO
MW: 210.6999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021397
CAS Number: 17380-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR