4-(Trifluoroacetyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Formyl-2,2,2-trifluoroacetophenone, 4-(2,2,2-trifluoroacetyl)benzaldehyde, 1-(4-Formylphenyl)-2,2,2-trifluoroethan-1-one

63628-500MG 63628-1G 63628-5G 63628-25G
77331-950EA 141.11 USD
77331-950 77344-856 77368-052 77389-394
4-(Trifluoroacetyl)benzaldehyde ≥97%
4-(Trifluoroacetyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547943
CAS Number: 86988-50-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR