Benzyl 2-isopropoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

30598-500MG 30598-1G 30598-5G 30598-25G
77325-124EA 201.88 USD
77325-124 77348-902 77361-462 77384-650
Benzyl 2-isopropoxynicotinate ≥97%
Benzyl 2-isopropoxynicotinate

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₃
MW: 271.311 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088169
CAS Number: 1826110-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR