Advanced MX Precision Balances
Supplier: Mettler Toledo
30665188 30665191 30665194 30665197
77323-112EA 8935.85 USD
77323-112 77323-116 77323-120 77323-124
Advanced MX Precision Balances
Balances and Scales
METTLER TOLEDO Advanced MX Precision Balances feature a large weighing pan, robust design, and precise weigh cell with FACT automatic internal adjustment.
  • SmartPan
  • Statistical data analysis
  • User management; Leveling assistant
  • StatusLight
  • 3 interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
  • Full metal housing with overload protection
The 7" intuitive touchscreen with user management provides automatic data logging, statistical analysis and easy access to 9 built-in weighing applications. Transfer data to a printer or computer forMore Product Information