AccuForge® Double Edge Blades, 400 Blade Dispenser
Supplier: Accutec Blades
AGBL-6008-0000
77322-454CS 1838.33 USD
77322-454
AccuForge® Double Edge Blades, 400 Blade Dispenser
Razors and Blades
AccuForge® Double Edge blades feature a 3-Facet edge for exacting, clean cuts.
  • 400 blade dispenser for high volume use
  • Easy access safety case
  • Clear cap for visibility
  • 3-Facet blade edge for increased sharpness
  • Degreased blade for reduced oil and pariculates
  • Made in USA
Paper-free dispenser is great for cleanroom use.
Precaución: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.