VWR® 96 Sample Purification System
Supplier: VWR International
77321-880 77321-882 77321-884 77321-886 77575-004
77321-880CS 510 USD
77321-880 77321-882 77321-884 77321-886 77575-004
VWR® 96 Sample Purification System
Automated Nucleic Acid Purification Systems
VWR® 96 Sample Purification System is a high throughput, automated system that improves the efficiency and reproducibility of magnetic bead-based purification protocols.
  • Compatible with all popular magnetic bead kits
  • On-board programming with a touch screen control panel
  • Process up to 96 samples in deep-well plates
  • 8 position carousel with 2 heating positions
  • Sample volume 50 to 1000 µl
Capable of processing up to 96 samples at one time, the VWR® 96 Sample purification system is easy to set up and program. Mixing, magnetic bead transfer, washing, andMore Product Information