(1R,2R)-(-)-2-Benzyloxycyclohexylamine 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-2-(Benzyloxy)cyclohexanamine

A140682-1G
103802-894EA 23 USD
103802-894 77316-002
(1R,2R)-(-)-2-Benzyloxycyclohexylamine 97%
(1R,2R)-(-)-2-Benzyloxycyclohexylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO
MW: 205.30 g/mol
Boiling Pt: 306.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01075752
CAS Number: 216394-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR