5,6-Dimethyl-1-indanone 96%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dimethyl-2,3-dihydro-1H-inden-1-one

A239475-1G
103811-294EA 44 USD
103811-294 103851-090 77304-162
5,6-Dimethyl-1-indanone 96%
5,6-Dimethyl-1-indanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MW: 160.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00666126
CAS Number: 16440-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR