Trimethyl orthobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Trimethyl orthobeznoate, (Trimethoxymethyl)benzene

A224259-100G A224259-10G A224259-500G A224259-25G A224259-5G A224259-1G
103819-094EA 5 USD
103819-094 103849-984 76786-092 76786-094 76786-096 77088-626 77088-628 77303-690
Trimethyl orthobenzoate 95%
Trimethyl orthobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.22
Boiling Pt: 206.3 °C (760 mm Hg)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008474
CAS Number: 707-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR