Fmoc-Lys-OH 97%

Supplier: Ambeed
Warning

A186089-100G A186089-10G A186089-5G A186089-1G A186089-25G
103815-910EA 6 USD
103815-910 103846-484 76998-842 76998-844 77084-132 77302-018 77302-020
Fmoc-Lys-OH 97%
Fmoc-Lys-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₄N₂O₄
MW: 368.44 g/mol
Boiling Pt: 607.6 °C (760 mm Hg)
Flash Pt: 321.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038539
CAS Number: 105047-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR