15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%

Supplier: Ambeed
Warning

Synonyms: 15-Crown-5, 1,4,7,10,13-Pentaoxacyclopentadecane

A142767-100G A142767-10G A142767-500G A142767-25G A142767-5G
77095-490EA 63 USD
77095-490 77095-492 77095-494 77299-310 77316-456
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₅
MW: 220.27 g/mol
Boiling Pt: 351
Melting Pt: -20
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005110
CAS Number: 33100-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR