3-Bromo-2-nitroaniline 98%

Supplier: Ambeed
Warning

A113957-100G A113957-10G A113957-25G A113957-1G A113957-5G
103798-326EA 9 USD
103798-326 103824-168 77094-118 77094-120 77094-122 77297-282 77297-284
3-Bromo-2-nitroaniline 98%
3-Bromo-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02
Boiling Pt: 298.4 °C (760 mm Hg)
Flash Pt: 134.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08063248
CAS Number: 7138-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR