4-Cyclohexyl-3-thiosemicarbazide 95%

Supplier: Ambeed
Danger

Synonyms: N-Cyclohexylhydrazinecarbothioamide

A549373-1G
77292-438EA 32.96 USD
77292-438
4-Cyclohexyl-3-thiosemicarbazide 95%
4-Cyclohexyl-3-thiosemicarbazide

Precaución: for Research Use Only

Formula: C₇H₁₅N₃S
MW: 173.2791 g/mol
MDL Number: MFCD00021318
CAS Number: 21198-18-5
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR