3-Nitrodibenzo[b,d]furan 95%

Supplier: Ambeed
Warning

A404186-250MG
77291-200EA 17 USD
77291-200
3-Nitrodibenzo[b,d]furan 95%
3-Nitrodibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₂H₇NO₃
MW: 213.19 g/mol
MDL Number: MFCD00092337
CAS Number: 5410-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR