3-Pyrimidin-5-ylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyrimidin-5-yl)benzoic acid

A184145-5G A184145-1G
77287-360EA 332.01 USD
77287-360 77748-196
3-Pyrimidin-5-ylbenzoic acid 97%
3-Pyrimidin-5-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₂
MW: 200.1900 g/mol
MDL Number: MFCD05864810
CAS Number: 852180-74-6
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR