4-Iodo-7-azaindole 97%

Supplier: Ambeed
Warning

A158640-10G A158640-1G A158640-250MG A158640-5G
77286-356EA 205.01 USD
77286-356 77286-358 77286-360 77286-362
4-Iodo-7-azaindole 97%
4-Iodo-7-azaindole

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MW: 244.0325 g/mol
MDL Number: MFCD08272234
CAS Number: 319474-34-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR