Thymidine (2'-deoxythymidine) 98%

Supplier: Ambeed
Warning

Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

A392461-25G A392461-1000G A392461-100G A392461-10G A392461-1G A392461-500G A392461-5G
76841-340EA 19 USD
76841-340 77278-634 77278-636 77278-638 77278-640 77278-642 77278-644
Thymidine (2'-deoxythymidine) 98%
Thymidine (2'-deoxythymidine)

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₅
MW: 242.23 g/mol
MDL Number: MFCD00006537
CAS Number: 50-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR