Methyl-3-fluoro-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-fluoro-4-hydroxybenzoate

A324688-10G A324688-1G A324688-250MG A324688-25G A324688-5G
77277-762EA 29 USD
77277-762 77277-764 77277-766 77277-768 77277-770
Methyl-3-fluoro-4-hydroxybenzoate 98%
Methyl-3-fluoro-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.14
MDL Number: MFCD00215838
CAS Number: 403-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR