(3-Chlorophenyl)acetone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Chlorophenyl)propan-2-one

A308645-10G A308645-1G A308645-5G
103814-566EA 18 USD
103814-566 103835-742 77277-466 77305-990 77305-992
(3-Chlorophenyl)acetone 95%
(3-Chlorophenyl)acetone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 236.542 °C (760 mm Hg)
Flash Pt: 116.264 °C
Storage Temperature: Ambient
MDL Number: MFCD00082872
CAS Number: 14123-60-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR