2-Methyl-2-pentenoic acid 99%

Supplier: Ambeed
Danger

A248702-10G A248702-25G A248702-5G
77276-264EA 6 USD
77276-264 77276-266 77276-268
2-Methyl-2-pentenoic acid 99%
2-Methyl-2-pentenoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14
MDL Number: MFCD00002655
CAS Number: 3142-72-1
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR