CORM-3 97%

Supplier: Ambeed
Warning

Synonyms: (OC-6-44)-Tricarbonylchloro(glycinato-κN,κO)ruthenium

A812133-1MG A812133-25MG A812133-50MG
77268-330EA 10 USD
77268-330 77268-332 77268-334
CORM-3 97%
CORM-3

Precaución: for Research Use Only

Formula: C₅H₄ClNO₅Ru
MW: 294.61 g/mol
MDL Number: MFCD07364048
CAS Number: 475473-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR