SANT-1 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine

A378069-10MG A378069-1MG A378069-25MG A378069-50MG A378069-5MG
77264-194EA 28 USD
77264-194 77264-196 77264-198 77264-200 77264-202
SANT-1 98%
SANT-1

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₅
MW: 373.5 g/mol
MDL Number: MFCD01827306
CAS Number: 304909-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR